Learn More
Sartorius™ Weighing Tables
Prevent damage to the surface of weights with these weighing tables.
Brand:  Sartorius™ YWT03
2330.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10529341
  • £2600.00 / Each
Explore more special offers
Description
Description
  • High density of the table's solid material provides excellent support for balances to meet the highest weighing requirements such as those stipulated for calibration of weights using mass comparators
  • Recommended if cleanliness of the weighing area is a major requirement
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Synthetic Stone
900 x 600 x 760 mm
300 kg
Weighing Table
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ Weighing Tables > Weighing Table; Material: Synthetic Stone