missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Welch™ Standard Duty WOB-L™ Piston Vacuum Pump: Model 2522
Shop All Welch Ilmvac Products
Click to view available options
Pump Type:
Vacuum Pump
Unit Size:
Each
Description
Includes
Includes convenient gauge read cut, pressure adjustment valve, protective separator jar, Euro (Schuko) plug
Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.
- Oil free dry pump powered by WOB-L piston
- Intake/Exhaust thread NPT: 1/4
Model 2522
- Free air displacement: CFM (L/min) at 60Hz: 0.76 (22)
- Ultimate pressure: 100 torr (133mbar)
- Maximum pressure: 100psig
- Maximum vacuum in Hg: 26
- Motor: 0.12 hp (93w)
- For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
- 50Hz units supplied with CE marking
Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box
Specifications
Specifications
| Horsepower | 0.12 hp |
| Shipping Weight (English) | 15 lb. |
| Shipping Weight (Metric) | 6.6 kg |
| Includes | Convenient gauge read cut, pressure adjustment valve, protective separator jar, Euro (Schuko) plug |
| Certifications/Compliance | CE |
| Electrical Requirements | 230 V 50 Hz |
| Model | 2522 |
| Weight (English) | 11.7 lb. |
| Weight (Metric) | 5.3 kg |
| For Use With | Vacuum filtration, vacuum oven, vacuum chamber/roughing, desiccation, aspiration, automation, cell culture |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering