missing translation for 'onlineSavingsMsg'
Learn More

Welch™ Standard Duty WOB-L™ Piston Vacuum Pump: Model 2561

For non-chemical applications

Brand:  Welch™ U2561C-50

1231.26 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 10095921


Includes: Includes convenient gauge read cut, pressure adjustment valve, protective separator jar
This item is not returnable. View return policy

Description

Description

Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.

  • Oil free dry pump powered by WOB-L piston
  • Intake/Exhaust thread NPT: 1/4

Model 2561

  • Free air displacement: CFM (L/min) at 60Hz: 2.1 (60); m2/hr at 50Hz: 3.0 (50)
  • Ultimate pressure: 5 torr (6.7mbar)
  • Maximum vacuum in Hg: 29.8
  • Motor: 0.3 hp (250w)

  • For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
  • 50Hz units supplied with CE marking

Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box

Specifications

Specifications

0.3 hp
9.7 kg
CE
2561
7.5 kg
Vacuum filtration, vacuum oven, vacuum chamber/roughing, desiccation, aspiration, automation, cell culture
10.5 in.
17.3 in.
6.5 in.
6.7 mbar
21.5 lb.
Convenient gauge read cut, pressure adjustment valve, protective separator jar, Euro (Schuko) plug
230 V 50 Hz
16.5 lb.
1.8 L/min.
50 Hz
26.7 cm
43.9 cm
Vacuum Pump
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers