missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellwash™ Microplate Washer (EU IVD/CE-marked)

Wash 96-well plates securely with this easy-to-use, reliable microplate-strip washer for routine ELISA applications.

£5070.32 - £5392.41

Specifications

Accuracy < 5% @ 300μL
Certifications/Compliance CE Marked
Connections USB
Wash/Waste Bottle Capacity 1
Bottles Wash Bottle (2 L), Waste Bottle (2 L)
View More Specs

Alert:

Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.


Products 2
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
15311095
View Documents
Thermo Scientific™
5165080
Wellwash 1x12 IVD model (Not available in USA) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15301095
View Documents
Thermo Scientific™
5165060
Wellwash 1x8 IVD model (Not available in USA) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.

Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.

Features:

  • Large color clear display and keypad for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Easy-to-use software requires minimal training for fast startup
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottle for security
  • Automatic prime feature assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizes if plate is present
  • Wellwash is designed to efficiently remove the excess liquid from the wells
  • Low residual volumes provide optimal washing performance and reliable assay results

Includes:

1x2L wash bottle, 1x2L waste bottle, 1x8L wash head, aerosol cover

Recommended for:

ELISA-based assays

Warranty: 1 year

Specifications

Specifications

< 5% @ 300μL
USB
Wash Bottle (2 L), Waste Bottle (2 L)
50 to 400 μL
<2 μL
50 to 1000 μL
ELISA-Based Assays
No
385 mm
345 cm
240 mm
17.6 lb.
100/240 V
CE Marked
1
< 3% (CV) @ 300μL (96-well plate)
96-Well Plates
8 kg
LCD
50/60 Hz
15.2 in.
13.6 in.
9.4 in.
No
100/240 V 50/60 Hz
5 to 100 mL
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.