Learn More
Thermo Scientific™ Wellwash™ Versa Microplate Washer
Wash 96- or 384-well plates securely with the Thermo Scientific™ Versa™ Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications.
£8690.00 - £8860.00
Specifications
Accuracy | < 5% @ 300μL, with n x 8 or n x 12 wash head; < 5% @ 250μL, with 2 x 8 cell wash head; < 5% @ 100μL, with 1 x 16 wash head |
---|---|
Certifications/Compliance | CE Marked |
Connections | USB |
Wash/Waste Bottle Capacity | 3 |
Bottles | Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
11555485
|
Thermo Scientific™
5165010 |
Wellwash Versa 2x8 |
£8690.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15184725
|
Thermo Scientific™
5165050 |
Wellwash Versa 2x12 |
£8860.00
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.
Simplify your ELISA workflow by using our eGuide to see our complete portfolio for all your ELISA needs.
Easy and convenient to use
- Large color display and keypad for easy, convenient use
- Clear display shows everything on one screen
- User interface is logic and intuitive, allowing quick protocol development
- Easy-to-use software requires minimal training for fast startup
Reliable and secure performance
- Unpressurized bottles are safe and secure to use
- Liquid level sensors are present in both wash and waste bottles
- Automatic rinse and prime features assure safe performance
- Aerosol cover prevents aerosolization of infectous diseases
- Plate sensor recognizeds if plate is present
Optimal washing performance
- Sweep mode guarantees an extremely low residual volume in the well, resulting in reliable assay results
- Washing performance is further optimized by adjustable parameters such as dispense and aspiration height, and aspiration speed
Added versatility
- A USB port for transferring protocols between several washers and for printing reports and log files
- Specially designed 2x8 cell wash head is available for cell washing
- Two 1x8 wash heads, two wash bottles and one rinse bottle, optional two 1x12 wash heads, 1x16 wash heads and 2x8 cell wash head offer increased flexibility
- Possibility to connect to automation systems
Includes:
2x2L wash bottles, 1x2L rinse bottle, 1x4L waste bottle, two 1x8 wash heads, aerosol cover
Recommended for:
ELISA-based assays, cell washing
Warranty: 1 year
Specifications
< 5% @ 300μL, with n x 8 or n x 12 wash head; < 5% @ 250μL, with 2 x 8 cell wash head; < 5% @ 100μL, with 1 x 16 wash head | |
USB | |
Wash Bottle (2 x 2 L), Rinse Bottle (1 x 2 L), Waste Bottle (1 x 4 L) | |
50 to 400 μL (96); 20 to 120 μL (384) | |
<2 μL | |
50 to 1000 μL (96); 20 to 300 μL (384) | |
LCD | |
50/60 Hz | |
15.2 in. | |
13.6 in. | |
9.4 in. | |
Yes (USB or RS-232C) | |
100/240 V 50/60 Hz | |
5 to 100 mL |
CE Marked | |
3 | |
< 3% (CV) @ 300μL, with n x 8 or n x 12 wash heads | |
96-well Plate, 384-well Plate | |
9 kg | |
5 to 100 mL | |
ELISA-Based Assays; Cell Washing | |
Yes | |
385 mm | |
345 mm | |
240 mm | |
20 lb. | |
100/240 V |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.