Learn More
Thermo Scientific™ Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
Brand: Thermo Scientific™ N12785
Description
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.Specifications
2 Pk. | |
Storage Bottle Cap | |
Wellwash Microplate Strip Washer |
Product Suggestions
Customers who viewed this item also viewed