missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, Plain

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.

Brand:  Cytiva 10400906

99.42 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 11305024

  • £146.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Filter Type: Plain
  • Pore Size: 0.8 μm
  • Autoclavable: Yes
  • Sterility: Non-sterile
  • Bubble Point: 1.3 bar
  • Flow Rate: 2.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Plain
ME 27
Non-sterile
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Plain
25 mm
1.3 bar
MCE
0.8 μm
2.8 sec./100 mL/12.5 cm⊃ (at Δp = 0.9 bar)
11.6 sec./100 mL (at Δp = 3 mbar)
0.98 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Flammable solid Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers