missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, Plain

Whatman™ (ME 27) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.

Brand:  Cytiva 10400914


 View more versions of this product

Product Code. 11325024

  • £143.15 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Material: Mixed Cellulose Ester
  • Material (Membrane): ME 27
  • Filter Type: Plain
  • Pore Size: 0.8 μm
  • Autoclavable: Yes
  • Sterility: Non-sterile
  • Bubble Point: 1.3 bar
  • Flow Rate: 2.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
  • Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
  • For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Plain
ME 27
Non-sterile
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Plain
50 mm
1.3 bar
MCE
0.8 μm
2.8 sec./100 mL/12.5 cm⊃ (at Δp = 0.9 bar)
11.6 sec./100 mL (at Δp = 3 mbar)
1.97 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ME27, 0.8 µm 50 mm, 100 pack

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers