missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 1.2μm Pore Size (ME 28) Mixed Cellulose Ester Membranes
Cytiva Whatman 1.2um Pore Size (ME 28) Mixed Cellulose Ester Membranes, Diameter Filter: 47mm, Pore Size: 0.2um
£137.00 - £424.00
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
11325034
|
Cytiva
10400821 |
100 mm |
£424.00
Pack of 50 |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
11335034
|
Cytiva
10400806 |
25 mm |
£137.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11345034
|
Cytiva
10400812 |
47 mm |
£164.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
11355034
|
Cytiva
10400814 |
50 mm |
£171.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Description
Cytiva Whatman 1.2um Pore Size (ME 28) Mixed Cellulose Ester Membranes, Diameter Filter: 47mm, Pore Size: 0.2um
Specifications
1.2 μm |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ||Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose