missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 3 μm Pore Size (ME 29) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 29) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
£129.00 - £171.00
Specifications
Autoclavable | Autoclavable |
---|---|
Filter Type | Plain |
Material | MCE |
Material (Membrane) | ME 29 |
Pore Size | 3 μm |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11385034
|
Cytiva
10400706 |
25 mm |
£129.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
11395034
|
Cytiva
10400712 |
47 mm |
£164.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
11305044
|
Cytiva
10400714 |
50 mm |
£171.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 29
- Filter Type: Plain
- Pore Size: 3 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Bubble Point: 1.3 bar
- Flow Rate: 1.2 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 6.7 sec./100 mL (at Δp = 3 mbar)
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
MCE | |
3 μm | |
1.2 sec./100 mL/12.5 cm² (at Δp = 0.9 bar) | |
6.7 sec./100 mL (at Δp = 3 mbar) |
Plain | |
ME 29 | |
Non-sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ||Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose