missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 4 and 13mm Puradisc Polyethersulfone Syringe Filters
For minimal adsorptive protein loss
£207.00 - £875.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Passes USP Class VI requirements for biosafety. |
Material | Polysulfone |
Sterility | Non-sterile |
For Use With (Application) | For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. |
Product Code | Brand | Pore Size | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Quantity | Sterility | Price | Quantity & Availability | |||
11304674
|
Cytiva
6782-1302 |
0.2 μm | 100/Pk. | Non-sterile |
£207.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11364664
|
Cytiva
6781-1304 |
0.45 μm | 500/Pk. | Non-sterile |
£875.00
Pack of 500 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||
11314674
|
Cytiva
6782-1304 |
0.45 μm | 100/Pk. | Non-sterile |
£210.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Description
Suitable for filtering serum, plasma and tissue culture solutions—as well as other protein-containing solutions where minimal adsorptive protein loss is required.
- Polypropylene housing
- Autoclavable
- Female Luer lock on inlet side; male slip Luer on outlet side
4mm dia. Filters
- For samples up to 2mL
- Filter area: 0.2cm2 Hold-up volume (with air purge) <7.5μL
- For samples up to 10mL
- Filter area: 1.3cm2
- Hold-up volume (after air purge) <25μL
Specifications
Autoclavable | |
Polysulfone | |
For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. | |
Female Luer Lok | |
Polypropylene | |
≤10 mL |
Passes USP Class VI requirements for biosafety. | |
Non-sterile | |
≤25 μL | |
Male Luer Slip | |
Syringe Filter | |
13 mm |