missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Acid-Treated Low-Metal TCLP Filters
Binder free filters
£99.50 - £342.00
Specifications
Description | GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action. |
---|---|
Format | Disc |
Material | Borosilicate Glass |
Particle Retention | 0.7 μm |
Shape | Circle |
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
11333135
|
Cytiva
1810-090 |
3.5 in. | 90 mm |
£99.50
Pack of 50 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11323135
|
Cytiva
1810-047 |
1.8 in. | 47 mm |
£125.00
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
11383125
|
Cytiva
1810-110 |
4.33 in. | 110 mm |
£139.00
Pack of 50 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11393125
|
Cytiva
1810-125 |
4.92 in. | 125 mm |
£180.00
Pack of 50 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11313135
|
Cytiva
1810-150 |
5.9 in. | 150 mm |
£342.00
Pack of 50 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11303135
|
Cytiva
1810-142 |
5.59 in. | 142 mm |
£230.00
Pack of 50 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
Description
Binder free filters
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm
- Resistant to acids and bases
- Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
- Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs
Compliance
Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.
Specifications
GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action. | |
Borosilicate Glass | |
Circle | |
TCLP Limits Test Filter |
Disc | |
0.7 μm | |
420 μm | |
60 mL/min |