Learn More
Cytiva Whatman AE98 Cellulose Nitrate Membranes
Recommended for the majority of routine applications
Brand:  Cytiva 10549916
Product Code. 15843704
  • £316.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured under strict cleanroom conditions
  • Can directly replace general puspose membrane filters
  • High protein binding
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AE98 Cellulose nitrate membranes
Cellulose Nitrate
Routine filtration applications
25mm x 50mm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • AE98 membrane, cellulose nitrate, 25mm x 50m
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose