missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Anotop™ 10mm Syringe Filters

Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.

Brand:  Cytiva 6809-1002

559.59 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : 75

Additional Details : Weight : 0.05040kg

 View more versions of this product

Product Code. 11384704

  • £916.00 / Pack of 50
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow

Specifications

Specifications

Single-use
0.02 μm
0.78 cm2
<20 μL
Hydrophilic
Male Luer Slip
Whatman™
45°C
Up to 10 mL
Anopore Inorganic Membrane (alumina matrix)
Non-sterile
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Polypropylene
Female Luer Lock
6.9bar (100psi)
50 Pack
Syringe Filter
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers