missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Autovial™ 5mL Syringeless Filters
Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.
£138.00 - £180.00
Specifications
Sterility | Non-sterile |
---|---|
Quantity | 50/Pk. |
Type | Syringeless Filter |
Volume (Metric) | ≤5 mL |
Diameter (Metric) | 13 mm |
Product Code | Brand | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
11879624
|
Cytiva
AV115NPEORG |
PTFE | 0.2 μm | 50/Pk. |
£180.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11393185
|
Cytiva
AV115NPUAQU |
PVDF | 0.45 μm | 50/Pk. |
£138.00
Pack of 50 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||
11373185
|
Cytiva
AV115NPUNYL |
Nylon | 0.45 μm | 50/Pk. |
£174.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11383185
|
Cytiva
AV115NPUORG |
PTFE | 0.45 μm | 50/Pk. |
£168.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
10389553
|
Cytiva
AV115UGMF |
Glass Microfiber | 0.45 μm | 50/Pk. |
£150.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Description
Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Remove particulates from samples up to 5mL in volume
- Filtration area: 1.7cm2
Glass Microfiber (GMF)
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Nylon (NYL)
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- Polytetrafluoroethylene (PTFE)
- Hydrophobic; for samples >50% organic solvents
- Polyvinylidene Fluoride (PVDF)
- Low protein binding; for aqueous samples
Specifications
Non-sterile | |
Syringeless Filter | |
13 mm |
50/Pk. | |
≤5 mL |