missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Autovial Syringeless Filters::Autovial 12::Without Prefilter
For removing particulates from potentially hazardous samples
Brand: Cytiva AV125NPUAQU
This item is not returnable.
View return policy
Description
Support stand protects slip-Luer tip, recessed to reduce chance of contamination. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial or fit a needle to its outlet for direct injection.
- Handle samples up to 12mL in volume
- No assembly required
- Replace syringe-coupled devices with single filter units for one-time use
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Available in PES and PVDF
Polyvinylidene Fluoride (PVDF)
Sample typr: Low protein binding. For aqueous samples, samples ≤60% organic solvent
Polyethersulfone (PES)
For aqueous samples, low protein binding
Specifications
Autovial 12 Syringeless Filter, no prefilter, 0.45 μm, PVDF filtration medium (50 pcs) | |
0.45 μm | |
Sample preparation, Difficult-to-filter samples, Quick filtration of samples, Low nonspecific protein binding membrane for samples in aqueous solutions and/or organic solvents. | |
Polypropylene (PP) | |
No | |
Up to 12 mL |
PVDF | |
3 cm2 | |
140 μL | |
Male slip luer | |
Syringeless Filter |