Learn More
Cytiva Whatman™ Binder-Free Glass Microfiber Prefiters, Grade GF/D
Functions as a prefilter when used with GF/B microfiber filters. Whatman™ Binder-Free Glass Microfiber Prefilters, Grade GF/D provide efficient membrane protection, especially during liquid scintillation counting.
£33.30 - £183.00
Specifications
Autoclavable | Autoclavable |
---|---|
Basis Weight | 121 g/m2 |
Description | GE’s Whatman Grade GF/D fine membrane filters are thick and offer good loading capacity. GF/D can also function as a prefilter when used with GF/B, and it comes in many sizes to fit most filter holders. |
Format | Disc |
Material | Borosilicate Glass |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11300374
|
Cytiva
1823-010 |
10 mm |
£41.50
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11390364
|
Cytiva
1823-007 |
7 mm |
£109.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11330374
|
Cytiva
1823-021 |
21 mm |
£33.30
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11340374
|
Cytiva
1823-024 |
24 mm |
£34.80
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11360594
|
Cytiva
1823-025 |
25 mm |
£76.75
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11390594
|
Cytiva
1823-090 |
90 mm |
£64.60
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11360374
|
Cytiva
1823-042 |
42.5 mm |
£43.35
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11370594
|
Cytiva
1823-047 |
47 mm |
£120.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11370374
|
Cytiva
1823-055 |
55 mm |
£68.15
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11380594
|
Cytiva
1823-070 |
70 mm |
£183.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11300604
|
Cytiva
1823-110 |
110 mm |
£44.60
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11310604
|
Cytiva
1823-125 |
125 mm |
£67.85
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11310374
|
Cytiva
1823-142 |
142 mm |
£90.50
Pack of 25 |
In Stock
Log in to see stock. |
||||||
11320604
|
Cytiva
1823-150 |
150 mm |
£125.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11330604
|
Cytiva
1823-257 |
257 mm |
£148.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11350374
|
Cytiva
1823-035 |
35 mm |
£40.50
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Functions as a prefilter when used with GF/B microfiber filters. Whatman™ Binder-Free Glass Microfiber Prefilters, Grade GF/D provide efficient membrane protection, especially during liquid scintillation counting.
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
- Thick and offer high loading capacity
- Function as a prefilter when used with GF/B.
- Fits most filter holders
- Nominal particle retention of 2.7μm
- Fast filtration speed
Specifications
Autoclavable | |
GE’s Whatman Grade GF/D fine membrane filters are thick and offer good loading capacity. GF/D can also function as a prefilter when used with GF/B, and it comes in many sizes to fit most filter holders. | |
Borosilicate Glass | |
2.6 s/100mL/in2 | |
Circle | |
500°C | |
675 μm | |
681 mL/min |
121 g/m2 | |
Disc | |
2.7 μm | |
Grade GF/D | |
932°F | |
0.02 in. | |
Prefilter |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.