missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Carbon-Cap Disposable Activated Carbon Capsules
Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.
£157.00 - £261.00
Specifications
For Use With (Application) | For water, chemical, and reagent purification |
---|---|
Housing Material | Polypropylene (PP) |
Max. Pressure | 4.1 bar (60 psi) |
Product Code | Brand | Filtration Area | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filtration Area | Price | Quantity & Availability | |||||
10526921
|
Cytiva
6704-1500 |
82,000 m2 |
£157.00
Each |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11314614
|
Cytiva
6704-7500 |
26000 m2 |
£261.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Filled with high-purity, high-efficiency, acid-washed, granular-activated carbon. Whatman™ Carbon Cap Disposable Activated Carbon Capsules are suitable for adsorption of organics from air and removal of color, organics and chlorine from water.
- Remove organic odors, color, and colloids from water and air
- Activated carbon plus a pleated HEPA filter media retains 99.97% of all particles 0.3μm
- Maximum housing pressure: 4.1 bar (60psi.)
- Carbon-Cap 75 offers 26,000m2 activated carbon surface area
- Carbon-Cap 150 offers 82,000m2 filter area equivalent
Applications:
For vacuum pump outlet removal of smoke and oil.
Specifications
For water, chemical, and reagent purification | |
Polypropylene (PP) | |
4.1 bar (60 psi) |