missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Cellulose Ester Membranes

Offers a high degree of internal surface area for greater adsorption of product

Brand:  Cytiva 10409814


Product Code. 11355014

  • £143.18 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Nontoxic grids that do not inhibit bacterial growth
  • Black plain and black gridded membranes have a mix of cellulose nitrate/cellulose
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane for high flow

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black/White Grid
Cellulose Ester
ME26/31
50 mm
Disc
0.6 μm
1.96 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • ||Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Whatman™ Cellulose Ester Membranes > Diameter: 50 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.