Learn More
Cytiva Whatman™ Cellulose Extraction Thimbles
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
£84.00 - £323.00
Specifications
Description 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
Material Cellulose
For Use With (Application) For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction
For Use With (Equipment) For automated extraction apparatuses
Max. Temperature (English) 248°F
View More Specs
Products 10
Product Code Brand Grade Length (Metric) Wall Thickness Diameter (Metric) Inside Price Quantity & Availability  
Product Code Brand Grade Length (Metric) Wall Thickness Diameter (Metric) Inside Price Quantity & Availability  
11373065
View Documents Promotion Details
Cytiva
10350215
603 60 mm 1.5 mm 25 mm
£102.00
Pack of 25
In Stock
to see stock.
Add to basket
11363065
View Documents Promotion Details
Cytiva
10350219
603 100 mm 1.5 mm 25 mm
£95.55
Pack of 25
In Stock
to see stock.
Add to basket
11323075
View Documents
Cytiva
10350226
603 80 mm 1.5 mm 28 mm
£96.60
Pack of 25
In Stock
to see stock.
Add to basket
11303075
View Documents Promotion Details
Cytiva
10350227
603 100 mm 1.5 mm 28 mm
£103.00
Pack of 25
In Stock
to see stock.
Add to basket
11310824
View Documents Promotion Details
Cytiva
10350240
603 80 mm 1.5 mm 33 mm
£121.00
Pack of 25
In Stock
to see stock.
Add to basket
11373075
View Documents Promotion Details
Cytiva
10350243
603 100 mm 1.5 mm 33 mm
£124.00
Pack of 25
In Stock
to see stock.
Add to basket
11383085
View Documents Promotion Details
Cytiva
10350265
603 123 mm 1.5 mm 41 mm
£192.00
Pack of 25
In Stock
to see stock.
Add to basket
11323095
View Documents
Cytiva
10350287
603 250 mm 2.5 mm 75 mm
£323.00
Pack of 25
In Stock
to see stock.
Add to basket
11323115
View Documents
Cytiva
10350437
603T 80 mm 1 mm 31 mm
£84.00
Pack of 25
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
11333095
View Documents
Cytiva
10350324
603 250 mm 3 mm 80 mm
£310.00
Pack of 25
Estimated Shipment: 27-08-2025
to see stock.
Add to basket
Description
Description
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
  • For all automated extraction apparatus
  • High-quality cellulose material
  • Dimensions matched exactly to thimble holders
Order Info
Extra allowance for wall thickness should be made when calculating external diameters
Specifications
Specifications
603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction
248°F
Extraction Thimble
Cellulose
For automated extraction apparatuses
130°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva Whatman™ Cellulose Extraction Thimbles