missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Cellulose Nitrate Membrane Filters

Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.

Brand:  Cytiva 10400114

Code : 69

Additional Details : CAS Number : 9004-70-0 Weight : 0.04500kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 11393135

  • £213.00 / 50g
Estimated Shipment: 07-08-2024
to see stock.

Description

Description

Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
Specifications

Specifications

Disc
8 μm
105 to 140 μm
50 mm
Cellulose Nitrate
100 Pack
1.96 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • AE99 membrane, cellulose nitrate

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : cellulose nitrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers