missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Filter Funnel Plate, Polypropylene

Use with Whatman™ 3-piece Filter Funnels. Whatman™ Filter Funnel Plate, Polypropylene is an optional extra plate recommended for most acids.

Brand:  Cytiva 1950-109

Code : 75

Additional Details : Weight : 0.04000kg

Product Code. 11300284

  • £78.02 / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • Recommended for most acids (except concentrated nitric acid and fuming sulfuric acid) at room temperature
  • Also suitable for most alcohols, glycols, ethers and ketones
  • Maximum working temperature: 100°C

Specifications

Specifications

Whatman 3-piece Filter Funnel
Recommended for most acids and also suitable for most alcohols, glycols, ethers and ketones
100°C
9 cm
Polypropylene
212°F
3.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers