missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Filter Funnel Replacement Reservoir

Filter Funnel Replacement Reservoirs for the 3-Piece Filter Funnel.

Brand:  Cytiva 1950-209

146.93 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Additional Details : Weight : 0.15300kg

 View more versions of this product

Product Code. 11380274

  • £177.00 / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Replacement filter funnel reservoir
Filter Funnel Reservoir
3.54 in.
1950 Series 3-piece Filter Funnels for glass microfiber filters
200 mL
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers