missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Filter Paper

Laboratory centrifuge accessory

Brand:  Cytiva 97039654

412.79 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : A0

Additional Details : Weight : 0.22000kg

Product Code. 11994556

  • £512.90 / Pack of 960
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

F319-04
960 per pack
44 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers