missing translation for 'onlineSavingsMsg'
Learn More
QIAGEN QIAcard FTA Elute Micro Product Code.: 11994416

QIAGEN QIAcard FTA Elute Micro

Product Code. 11994416
100 Pk., Pack of 100
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 11994416

Brand: QIAGEN WB120410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

  • FTA Elute technology for convenient water based elution of nucleic acids.
  • QIAcard FTA Elute Micro format with 4 spot area (spot volume 12 to 40 ÎĽL) to collect, stabilize, process, transport, and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

Specifications

Content And Storage Room Temperature
Description Collect, process, transport and store colored biological samples, such as blood samples with Non-Indicating FTA Elute Micro Cards.
Sample Type Blood
For Use With (Application) Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
No. of Samples 4
Product Type FTA Elute Micro Card
Quantity 100 Pk.
Salt Form Chaotropic
Sample Volume 40 ÎĽL
Test Time 15 to 30 min
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
QIAGEN QIAcard FTA Elute Micro >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.