Learn More
QIAGEN QIAcard FTA CloneSaver
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. 96 sample areas per card.
Brand:  QIAGEN WB120028
Product Code. 11989424
  • £180.00 / Pack of 5
Explore more special offers
Description
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. 96 sample areas per card.
CloneSaver Card is designed for the collection, long-term storage and purification of plasmid and BAC DNA from bacterial clones in a 96-well format.
Specifications
Specifications
Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96-well format.
Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96 well format.
1 Pk.
96-well Plate
CloneSaver Card
5 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
SDS
Documents
Documents
Certificates
View accessory products