Learn More
QIAGEN QIAcard FTA Wash Buffer
For contamination-free collection, storage and transport of DNA samples.
Brand: QIAGEN WB120204
This item is not returnable.
View return policy
Description
For contamination-free collection, storage and transport of DNA samples.
QIAcard FTA Wash Buffer is used for purifying nucleic acids stored on FTA cards.
Specifications
QIAcard FTA cards, indicating FTA cards and EasiCollect device | |
Rapid purification of DNA from blood, cells, tissue, yeast, bacteria | |
500 mL |
Product Content Correction
Product Title
QIAGEN QIAcard FTA Wash Buffer > 500 mL
Spot an opportunity for improvement?Share a Content Correction