Learn More
Cytiva Whatman™ Glass Fiber Tapes, Grade HGF61
For continuous air monitoring, compatible with air samplers from multiple manufactures globally
Brand: Cytiva 1830-10091
Additional Details : Weight : 0.61000kg
Description
- Provides strength, efficiency, reliability and consistency of data quality in continuous air monitoring
- Meets regulatory requirements for continuous air particulate sampling globally
- Suitable for PM1, PM2.5, PM10 and TSP sampling and for beta-attenuation method
- Provides excellent consistency of data quality in air sampling
- Effective in both humid and dry environments
- Compatable with most continuous air sampling devices
- Retains 99.99% of air particulate at 0.3 μm
Specifications
54 g/m2 | |
Continuous Air Monitoring in High Humidity Environments | |
1 Reel | |
Continuous Air Monitor Tape | |
30 mm |
Glass Microfiber with Binder | |
Grade HGF61 | |
285 μm | |
40 m |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.