Learn More
Cytiva Whatman™ Glass Microfiber Extraction Thimbles, Grade 603G
Flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. Whatman™ Glass Microfiber Extraction Thimbles are designed for applications including analysis of hot and acidic gases, analysis of pesticide residues, determination of oil/fat content of foods, and analysis of oil and grease in solid wastes.
£166.00 - £298.00
Specifications
Description | GE’s Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. |
---|---|
Material | Borosilicate Glass Fibers |
For Use With (Application) | Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes. |
Grade | 603G |
For Use With (Equipment) | Soxhlet extractor |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inside | Wall Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inside | Wall Thickness | Price | Quantity & Availability | ||||
11363095
|
Cytiva
10371005 |
50 mm | 16 mm | 1 mm |
£166.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11303105
|
Cytiva
10371019 |
100 mm | 25 mm | 1.5 mm |
£186.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11373095
|
Cytiva
10371007 |
90 mm | 19 mm | 1 mm |
£196.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11383095
|
Cytiva
10371011 |
80 mm | 22 mm | 1.5 mm |
£168.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11353105
|
Cytiva
10371036 |
100 mm | 30 mm | 1.5 mm |
£236.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11343105
|
Cytiva
10371025 |
60 mm | 28 mm | 1.5 mm |
£186.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11383105
|
Cytiva
10371042 |
94 mm | 33 mm | 1.5 mm |
£201.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11363105
|
Cytiva
10371043 |
100 mm | 33 mm | 1.5 mm |
£200.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11393105
|
Cytiva
10371055 |
150 mm | 35 mm | 1.5 mm |
£298.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11393095
|
Cytiva
10371114 |
68 mm | 23.8 mm | 1.5 mm |
£168.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11373105
|
Cytiva
10371045 |
118 mm | 33 mm | 1.5 mm |
£253.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11353095
|
Cytiva
10371103 |
38 mm | 10 mm | 1 mm |
£168.00
Pack of 25 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
Description
- Resistant to temperatures up to 500°C (932°F)
- Manufactured with walls as thin as 1–1.5mm
- Standard glass Soxhlet thimble applications
- 603G borosilicate glass fiber thimble with inorganic binder
Specifications
GE’s Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. | |
Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes. | |
Soxhlet extractor | |
Extraction Thimble |
Borosilicate Glass Fibers | |
603G | |
500°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.