missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Glass Microfiber Filter Paper Grade 6

Captures tiny particles without becoming weak. Whatman™ Glass Microfiber Filter Paper Grade 6 may also be used for determination of filterable substances and residue upon ignition or analysis of aggressive media.

Brand:  Cytiva 10370004

111.40 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : A0

Additional Details : Weight : 0.05580kg


 View more versions of this product

Product Code. 11305264

  • £123.00 / Pack of 100
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

Suitable for water and wastewater analysis, with extremely fine particle retention and added strength.
    • Sub micron filter retention efficiency to capture extremely small particles
    • Inorganic binder for increased strength
    • Temperature-resistant up to 500°C
    • Can be used as a pre-filter for dense samples
Specifications

Specifications

Disc
<1 μm
Slow
100 Pack
2.75 in.
Borosilicate Glass
40 s/100mL/in2
GF 6
350 μm
70 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FILTER GLASS CIRCLE

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers