missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper

17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.

£77.85 - £1210.00

Specifications

Description 17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
Flow Rate 190 mm/30 min.
Film Thickness 0.92 mm
For Use With (Application) Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Grade Grade 17 Chr
View More Specs

Products 3
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
10622245
View Documents
Cytiva
3017-8793
22 cm 2.5 cm Sheet
£77.85
Pack of 100
In Stock
to see stock.
Add to basket
 
11909197
View Documents
Cytiva
3017-621
30 cm 2.5 cm Roll
£88.45
Each
In Stock
to see stock.
Add to basket
 
11308854
View Documents
Cytiva
3017-917
57 cm 46 cm Sheet
£1210.00
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
Specifications

Specifications

17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
0.92 mm
Grade 17 Chr
190 mm/30 min.
Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Cellulose
Videos
SDS
Documents

Documents

Product Certifications