missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 32 Binder-Free Glass Fiber Filter

Brand:  Cytiva 10372968

Code : A0

Additional Details : Weight : 0.23000kg

Product Code. 11318914

  • £203.00 / Pack of 50
Estimated Shipment: 12-12-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Glass Fiber
1 μm
32
Sheet
Slow
10.1 x 30.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers