missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 556 Drypads

Product Code. 11919337
Click to view available options
Dimensions (L x W):
37 x 100 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11919337

Brand: Cytiva 10310992

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Absorbent paper strips for removal of excess liquids

  • Absorbent paper strips for removal of excess liquids, e.g., from microscope specimens
TRUSTED_SUSTAINABILITY

Specifications

Description 556 Drypads 37x100 mm 100/Pk
Material Cellulose
For Use With (Application) Absorbent paper strips for the removal of excess liquids, e.g., from microscope specimens.
Grade Grade 556
Quantity 100 Pk.
Type Drypad
Dimensions (L x W) 37 x 100 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.