Learn More
Cytiva Whatman™ Grade 603 Cellulose Extraction Thimbles
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
£83.35 - £259.00
Specifications
Description | 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. |
---|---|
Material | Cellulose |
For Use With (Application) | To analyze fats and extract organic compounds |
Grade | 603 |
Max. Temperature (English) | 266°F |
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
11383065
|
Cytiva
10350217 |
25 mm | 80 mm | 1.5 mm |
£98.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11313075
|
Cytiva
10350225 |
28 mm | 60 mm | 1.5 mm |
£83.35
Pack of 25 |
In Stock
Log in to see stock. |
||||
11343095
|
Cytiva
10350220 |
26 mm | 60 mm | 1.5 mm |
£91.55
Pack of 25 |
In Stock
Log in to see stock. |
||||
11393065
|
Cytiva
10350223 |
27 mm | 80 mm | 1.5 mm |
£83.35
Pack of 25 |
In Stock
Log in to see stock. |
||||
11343075
|
Cytiva
10350234 |
30 mm | 80 mm | 1.5 mm |
£88.65
Pack of 25 |
In Stock
Log in to see stock. |
||||
11313085
|
Cytiva
10350238 |
33 mm | 60 mm | 1.5 mm |
£84.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10237830
|
Cytiva
10350236 |
30 mm | 100 mm | 1.5 mm |
£108.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10232031
|
Cytiva
10350245 |
33 mm | 118 mm | 1.5 mm |
£150.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11323085
|
Cytiva
10350241 |
33 mm | 90 mm | 1.5 mm |
£103.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11393075
|
Cytiva
10350247 |
33 mm | 130 mm | 1.5 mm |
£123.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11303085
|
Cytiva
10350250 |
33 mm | 205 mm | 1.5 mm |
£183.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11343085
|
Cytiva
10350252 |
34 mm | 130 mm | 1.5 mm |
£123.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11373085
|
Cytiva
10350261 |
40 mm | 85 mm | 1.5 mm |
£126.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11363085
|
Cytiva
10350255 |
35 mm | 150 mm | 1.5 mm |
£148.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11303095
|
Cytiva
10350273 |
48 mm | 145 mm | 2 mm |
£187.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11343065
|
Cytiva
10350306 |
22 mm | 60 mm | 2 mm |
£121.00
Pack of 25 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||||
11393085
|
Cytiva
10350275 |
44 mm | 230 mm | 2 mm |
£259.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11333085
|
Cytiva
10350242 |
33 mm | 94 mm | 1.5 mm |
£124.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
11313095
|
Cytiva
10350274 |
48 mm | 200 mm | 2 mm |
£220.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10097740
|
Cytiva
10350211 |
22 mm | 80 mm | 1.5 mm |
£137.00
Pack of 25 |
In Stock
Log in to see stock. ![]() |
||||
Description
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Pure Cellulose Thimbles
- Made from pure cotton cellulose
- For extraction of pesticides from environmental samples, fat analysis in the food industry
- Single-thickness thimbles have wall thickness of approximately 1mm
- Double-thickness thimbles have ∽2mm wall thickness
- For high-temperature extractions up to 550°C (1022°F). Also for using solvents incompatible with cellulose thimbles
Specifications
603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
To analyze fats and extract organic compounds | |
266°F | |
Extraction Thimble |
Cellulose | |
603 | |
130°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.