Learn More
Cytiva Whatman Grade GF/C Glass Microfiber Filters, Binder Free, Ready to Use
Grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity, and support total suspended solids analysis.
Brand: Cytiva 3822-090
152.22 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.35000kg
Description
- Good flow rate and loading capacity
- Fine nominal particle retention of 1.2μm
- Binder free
- Applications include total suspended solids analysis, industrial waste analysis and potable water
- GF/C RTU pre-treats traditional GF/C filter with washing, drying, cooling (in a desiccator), and weighing steps aligned to EN 872
- Certified weight loss of less than 0.017mg/cm2 after preparation
- Each RTU filter is placed in an individually barcoded aluminum pan that notes the filter weight
Specifications
6.7s/100mL/in2 | |
Borosilicate Glass | |
1.2 μm | |
Grade GF/C | |
550°C | |
RTU Filter, pre-weighed | |
9.0 cm |
GE’s ready-to-use Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 μm. | |
1.2 μm | |
Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is | |
100 Pieces | |
260 μm | |
105 mL/min |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.