Learn More
QIAGEN QIAcard FTA Elute Indicating Micro
For collection, stabilization, processing, transport and archiving of biological samples. 1 sample area per card.
Brand:  QIAGEN WB120412
 View more versions of this product
Product Code. 12445432
  • £145.00 / Pack of 25
Explore more special offers
Description
Description
  • FTA Elute technology for convenient water based elution of nucleic acids.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
Room Temperature
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
Indicating FTA Elute Micro Card
40 μL
Collect, process, transport, and store colorless biological samples, such as buccal or saliva samples. With FTA Elute, proteins remain tightly bound while DNA is eluted from the matrix.
Inert dye
25 Pk.
15 to 30 min
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
QIAGEN QIAcard FTA Elute Indicating Micro > 25 Pk.