Learn More
Cytiva Whatman™ Lead Acetate Test Paper Dispenser
Dispenser for rapid qualitative test paper to detect hydrogen sulfide
Brand: Cytiva 2602-501A
19.17 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 69
Additional Details : Weight : 0.01980kg
Description
Lead Acetate Test Paper Detections
- As little as 5ppm of atmospheric or gas stream H2S
- As little as 4ppm H202 by preblackening paper with H2S
Specifications
For rapid qualitative test paper to detect hydrogen sulfide, detect ∼5ppm of H2S in atmosphere or gas stream | |
Each | |
Lead Acetate Test Paper Dispenser |
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, lead(2+) salt, trihydrate