missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ MicroPlus Membranes
Click to view available options
Diameter (Metric):
47 mm
50 mm
Color:
Black
Green
White
Grid Color:
Black
White
Filter Code:
MicroPlus-21 STL
MicroPlus-21ST
MicroPlus-31STL
MicroPlus-41STL
Unit Size:
Pack of 100
Pack of 400
Description
The membrane filtration (MF) method is excellent for the determination and enumeration of microbial contaminants.
- Special membrane filters made from cellulose nitrate developed for microbiological quality control
- High mechanical strength
- High flow rate
- Sterile, individually packed
- All membrane filters have a high-contrast grid (3.1 mm)
- Type STL in dispenser boxes for easy removal and secure handling with the Membrane-Butler
Specifications
Specifications
| Color | Green |
| For Use With (Equipment) | Membrane Butler |
| Material (Membrane) | Cellulose Nitrate |
| Pore Size | 0.45 μm |
| Sterility | Sterile |
| Filter Code | MicroPlus-41STL |
| For Use With (Application) | Microbiological quality control |
| Grid Color | Black |
| Size (Metric) Grid | 31mm |
| Surface Type | Grid |
| Show More |
Safety and Handling
Hazard Category
- Flammable solid Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction