missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

Brand:  Cytiva UN203NPEAQU

229.12 GBP valid until 2024-12-31
Use promo code "21934" to get your promotional price.



Code : 69

Additional Details : Weight : 1.58760kg

 View more versions of this product

Product Code. 11388734

  • £269.00 / Pack of 100
Estimated Shipment: 14-11-2024
to see stock.
Add to basket

Description

Description

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.

  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 μm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
Specifications

Specifications

Translucent
Single-use
PTFE-coated Silicone Rubber Septa
0.2 μm
Polypropylene (PP)
100/Pk.
50°C
0.4 mL
Equivalent in size to 12 x 32mm vials
Autosampler, Compressor
PVDF
Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Vial with incorporated filtration membrane, plunger and cap/septa
0.6 bar / 8 psi (to compress)
Syringeless Filter
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions