missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Mixed Cellulose Ester Membrane Filters

Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.

Brand:  Cytiva 10407334

Code : 53

Additional Details : Weight : 0.04300kg


 View more versions of this product

Product Code. 11315384

  • £476.00 / Pack of 400
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • Sterile options available for critical applications
  • Contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
Specifications

Specifications

Autoclavable
Membrane Butler
0.45 μm
To quantitatively evaluate microorganism counts
Hydrophilic
400
0.31cm
266°F
Membrane
50 mm
Black
MCE
Sterile
White
ME25
Medium
Grid
130°C
1.96 in.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers