Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membrane Filters
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
Brand: Cytiva 10407372
Code : 53
Additional Details : Weight : 0.04300kg
Description
- Sterile options available for critical applications
- Contrast for easier particle detection
- Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
- Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
- Higher dirt loading capacity
- Biologically inert with good thermal stability
- No surfactants to contaminate samples
- Uniform microporous structure of membrane gives high flow rates
Specifications
Autoclavable | |
Membrane Butler | |
0.45 μm | |
To quantitatively evaluate microorganism counts | |
Hydrophilic | |
400 | |
0.31cm | |
266°F | |
Membrane | |
50 mm |
Green | |
MCE | |
Sterile | |
Balck | |
ME25 | |
Medium | |
Grid | |
130°C | |
1.96 in. |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose