Learn More
Cytiva Mixed Cellulose Ester Membranes, WME, Circles
Smoother, more uniform surface than pure nitrocellulose filters
Brand: Cytiva 7153-104
Code : A0
Additional Details : Weight : 0.17100kg
Description
- Sterile for critical applications
- Excellent contrast for easier particle detection
- Nontoxic grids ensure sample integrity
- Black gridded membranes
- Mix of cellulose nitrate and cellulose acetate
- High degree of internal surface area for greater adsorption of product
- Higher dirt loading capacity
- Biologically inert with good thermal stability
- No surfactants to contaminate samples
- Uniform microporous structure of membrane gives high flow rates
- Thermally stable
- Clarification or sterilization of aqueous solutions
- Cytology
- Air monitoring
- HPLC samples (aqueous)
- Virus concentration
- Particulate analysis
- Biological assays
- Food microbiology including enumeration of E. coli in foods
- Bacteriological studies
- Particle counting from liquids and aerosols
- Yeasts and molds
Specifications
Black | |
0.45 μm | |
Yes | |
Gridded (White grid), with pad |
MCE | |
Sterile | |
100 | |
47 mm |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose