Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Brand: Cytiva 7141-114
111.01 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
Order Info
Autoclavable pack has 10 envelopes of 10 filters/pads for sterilization in the microbiological laboratory.
Specifications
Autoclavable | |
Mixed Cellulose Ester | |
0.45 μm | |
Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions | |
Interleaving paper | |
Medium | |
3.1mm | |
266°F | |
1.85 in. |
White | |
WME | |
Sterile | |
Black | |
Individually Packed | |
100 Pack | |
Grid | |
130°C | |
47 mm |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose