Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Marca: Cytiva 7141-204
Code : 75
Dettagli aggiuntivi : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Descrizione
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
Specifica
Autoclavable | |
Mixed Cellulose Ester | |
0.45 μm | |
Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions | |
Interleaving paper | |
Medium | |
3.1mm | |
266°F | |
1.85 in. |
White | |
WME | |
Non-sterile | |
Black | |
Autoclave Pack | |
100 Pack | |
Grid | |
130°C | |
47 mm |
Sicurezza e movimentazione
- Flammable solid Category 1