Learn More
Cytiva Whatman™ Nuclepore Hydrophilic Membrane
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
Brand:  Cytiva 10418501
65.50 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16615675
  • £71.70 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to make it hydrophilic
  • Low extractables minimizing filtration sample contamination risk
  • High chemical resistance
  • Good thermal stability
  • Low, consistent ash
  • Low tare weights
  • Smooth, flat membrane surface for high particle visibility
Applications:
  • Bioassays
  • Cell biology
  • Environmental analysis
  • Microscopy
  • Cytology
  • Parasitology
  • Certain air and oceanography analyses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate
Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses
Nuclepore hydrophilic membrane
12 μm
100 Pk.
13 mm