Learn More
Cytiva Whatman™ Nuclepore Hydrophilic Membrane
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
Brand: Cytiva 10418312
This item is not returnable.
View return policy
Description
- Track-etched for highly defined pore size and high pore density
- Treated with PVP to make it hydrophilic
- Low extractables minimizing filtration sample contamination risk
- High chemical resistance
- Good thermal stability
- Low, consistent ash
- Low tare weights
- Smooth, flat membrane surface for high particle visibility
Applications:
- Bioassays
- Cell biology
- Environmental analysis
- Microscopy
- Cytology
- Parasitology
- Certain air and oceanography analyses
Specifications
Polycarbonate | |
Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses | |
Nuclepore hydrophilic membrane |
3 μm | |
100 Pk. | |
47 mm |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Cytiva Whatman™ Nuclepore Hydrophilic Membrane > Pore Size: 3 μm, Diameter: 47 mm
Spot an opportunity for improvement?Share a Content Correction