Learn More
Cytiva Whatman™ Nuclepore Hydrophilic Membrane
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
Brand:  Cytiva 10418401
 View more versions of this product
Product Code. 16675665
  • £73.90 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to make it hydrophilic
  • Low extractables minimizing filtration sample contamination risk
  • High chemical resistance
  • Good thermal stability
  • Low, consistent ash
  • Low tare weights
  • Smooth, flat membrane surface for high particle visibility
Applications:
  • Bioassays
  • Cell biology
  • Environmental analysis
  • Microscopy
  • Cytology
  • Parasitology
  • Certain air and oceanography analyses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate
Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses
Nuclepore hydrophilic membrane
10 μm
100 Pk.
13 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Nuclepore Hydrophilic Membrane > Pore Size: 10 μm, Diameter: 13 mm