Learn More
Cytiva Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Suitable for a wide range of sample types and applications including bioassays, cell biology, and environment analysis
Brand: Cytiva 10417406
70.98 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- A highly defined pore size cutoff in the sub-micron range enables particle deformity measurement and the capture of extremely small particles
- Membranes are available in a range of pores sizes, diameters, and sheet formats
- Track-etched for highly defined pore size and high pore density
- Treated with polyvinylpyrrolidone (PVP) to render it hydrophilic
- Low extractables, minimizing filtration sample contamination risk
- High chemical resistance and good thermal stability for a wide range of sample types
- Low consistent ash and tare weights
- Smooth flat membrane surface for high particle visibility
Specifications
5 μm | |
Polycarbonate Film Track-Etched (PFTE) | |
Nucleopore Hydrophilic Membrane |
Microscopy, Cytology, Parasitology, as Well as Certain Air and Oceanography Analyses | |
100/Pk. | |
25 mm |