missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nylon Membranes

Product Code. p-112196
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.5 μm
0.8 μm
Diameter (Metric):
13 mm
25 mm
47 mm
90 mm
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 11384864

Brand: Cytiva 7402009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

 

  • Pure nylon membranes
  • Tear-resistant
  • Filter many organic solvents
  • Autoclavable up to 135°C
  • Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
  • Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

 

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Nylon
Pore Size 0.2 μm
For Use With (Application) For filtering aqueous solutions and most organic solvents
Hydrophobicity Hydrophilic
Quantity 50/Pk.
Diameter (Metric) 90 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.