Learn More
Cytiva Whatman™ pH Indicators
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Brand: Cytiva 10362000
16.95 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
Specifications
0 to 14 | |
Color comparison chart | |
Indicator Paper |
Strip | |
100 Pack |